Would you like to log in again?
MS 3 Digital Vortex Mixers, IKA® Works
Supplier: IKA
0003319000
89209-860EA 1256.51 USD InStock
89209-860
MS 3 Digital Vortex Mixers, IKA® Works
Shakers and Mixers Vortex Mixers
Compact, universal small shaker suitable for shaking tasks with all small vessels and microtiter plates.
  • Wide range of attachment with detection
  • Timer with countdown function
  • Continuous or touch operation
  • Units are stable in all speed ranges and features sturdy zinc die cast casing
Two operating modes, In Mode A (safe mode with attachment detection), the maximum speed of 3000 rpm is only reached with the standard attachment in touch mode. When using other attachments the speed is limited to 1300 rpm. In Mode B (without attachment detection) a speed of 3000 rpm is possible with all attachments.
Certifications: Units meet IP 21 protection class according to DIN EN 60529.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty