Would you like to log in again?
WHEATON® Peristaltic Pumps, Dispensing Pumps and Tubing
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
W375025 374320 W375040-B I054348 W374709 W374730 W374706 W374701 W374733 374305 W375023 W375024 374321 W375040-D W375040-J W375030-G W375040-A 374304 W375040-G W374707 W374737 W374708 I054346 W375030-D W375040-C W375030-F 374326 W374700 W374705 W375030-C I054342 W375030-J 374308 W375030-A W375022 W375040-F W375030-B W374738 W374702 374324 W374703 W374734 374322 W374731 374328 374306 W374735 I054325 I050694 W374704
25485-031EA 43.8 USD InStock
25485-031 25485-030 14006-832 14006-834 89194-330 89194-334 89212-200 89194-332 89194-338 89194-336 89212-208 25485-038 25485-037 25485-035 89212-204 89212-202 14231-588 16171-481 14231-585 89212-210 14231-582 89212-218 89212-216 89212-212 14231-318 14231-594 89194-352 89194-350 14231-590 89194-356 89212-188 89194-354 89212-186 89212-184 89212-182 14230-632 14230-630 89212-192 89212-190 89194-342 89194-340 89194-346 89194-344 89212-198 89212-196 25485-029 89212-194 89194-348 14006-836 14006-838
WHEATON® Peristaltic Pumps, Dispensing Pumps and Tubing
Pumps Peristaltic Pumps
Wheaton peristaltic pumps are used for dispensing and general liquid handling purposes
The pumps are delivered ready to use with tubing kits included. The pumps can be programmed to dispense measured aliquots into culture plates, vials, tubes or bottles. The Omnispense units have additional features including speed ramping and totalizer/flow mode.
Ordering information: Includes complete pump, instruction manual and tubing kits. Works with 2.3mm thick tubing. Additional pump heads and foot switches available.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty