Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SpectraMax® Plus 384 Absorbance Plate Reader, Molecular Devices
Supplier: Molecular Devices
0310-3258 PLUS-384-SC
89212-396EA 52140.43 USD InStock
89212-396 10067-760
SpectraMax® Plus 384 Absorbance Plate Reader, Molecular Devices
Microplate Readers
This UV-visible absorbance microplate reader provides ultrafast, full spectral range detection for cuvettes, 96-well, and 384-well microplates, with the only temperature-independent method for pathlength correction.
  • Temperature control
  • Shaking capabilities
  • Multichannel optical design
  • Eliminates error
Temperature control and shaking come standard.
The patented multichannel optical design of the SpectraMax® Plus384 Absorbance Microplate Reader mimics a dual beam spectrophotometer, measuring each sample in the plate directly, eliminating measurement error due to variations in light output between optical fibers on other instruments for more reliable data reporting. The nine-channel system delivers both high precision and speed when reading cuvettes, 96-well, and 384-well microplates for endpoint measurements, kinetic assays, and spectral scans. The optical system of SpectraMax Plus384 Microplate Reader is built around a monochromator, which allows for selection of up to six wavelengths at a time for absorbance detection in the UV-visible wavelength range (190 nm - 1000 nm). Narrow 2nm bandwidths deliver increased measurement accuracy and linearity over the widest range of assays, including ELISAs in the visible range or DNA quantitation in the UV range.
The SpectraMax® Plus can read as low as 2μL with the μMax Low Volume Plate. The SpectraMax® also uses patented PathCheck® Pathlength Measurement Technology: the only temperature independent pathlength correction for microplates.
Ordering information: Includes current version of SoftMax Pro Software. Additional models are available. Additional accessories are available including the µMax Low Volume Plate, standard validation tools, and 21 CFR Part 11 Compliance tools. Standard 12-month warranty included. Purchase of Additional 1 Year Service Plan includes: MRC Essential Service Plan, including parts, labor, and shipping/handling costs for emergency repairs for one year at the Molecular Devices Repair Center. Priority repair scheduling, along with unlimited technical support and loaner instrument (if available), are provided with this service option.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty