Would you like to log in again?
CARTRIDGE MS-PP11 15ML PK50
Supplier: Agela Technologies
Cartridge Ms-Pp11 15Ml 50/Pack^ Clean-Up tube, suitable for vegetable and fruit with pigment; suitable for en methord 15662;
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE MS-PP11 15ML PK50 89213-682 MS-PP1511 Pack of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty