Would you like to log in again?
SPE Vacuum Manifolds, Bonna-Agela Technologies
Supplier: Agela Technologies
VM24 VM12
89213-858EA 3690.55 USD InStock
89213-858 89213-860
SPE Vacuum Manifolds, Bonna-Agela Technologies
Manifolds
SPE vacuum manifolds are ideal for SPE sample preparation, filtration and elution.
  • Provides consistent extraction and filtration results
  • Solvent resistant and autoclavable lid
  • Adjustable racks will accommodate a variety of sample collection vessels
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty