Would you like to log in again?
VWR® CryoPro® Plastic Storage Boxes
Supplier: VWR
VWR® CryoPro®
04A1-25M-PB 04A1-81-PB 04A1-81-PB-T 04A1-100-PB
89214-749CS 637.16 USD InStock
89214-749 89214-746 89214-762 89214-760
VWR® CryoPro® Plastic Storage Boxes
Boxes Cryoboxes
Polycarbonate construction for use in mechanical and LN2 freezers.
  • Grid format for lids
  • Boxes feature drain slots for safe liquid nitrogen storage
Lids come printed with 25, 81, and 100 cell grids to assist with inventory control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty