Would you like to log in again?
VWR® Cryogenic Fiberboard Freezer Box
Supplier: Avantor
04A1-02M-25G 04A1-02-100G 04A1-02-GEN 04A1-03-GEN 04A1-02-81-G
89214-758CS 265.36 USD InStock
89214-758 89214-752 89214-750 89214-756 89214-754
VWR® Cryogenic Fiberboard Freezer Box
Boxes Cryoboxes
Fiberboard boxes can withstand LN₂ storage and do not deteriorate wtihin the LN₂, breakdown can occur if the boxes are removed from the LN₂ and left to condensate in ambient temperatures.
  • Multiple heights of boxes to accommodate a variety of heights of vials/tubes (purchase divider wtih empty box)
  • Printed alignment arrows (fiberboard boxes with grids only)
  • Drain slots for LN₂ drainage (fiberboard boxes with with grids only)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty