Would you like to log in again?
Adapter, Distillation, Ace Glass Incorporated
Supplier: Ace Glass
5299-12 5299-07 5299-10 5299-01 5299-16 5299-03
89214-890EA 863.81 USD InStock
89214-890 89214-880 89214-884 89214-882 89214-888 89214-886
Adapter, Distillation, Ace Glass Incorporated
Distillation Components Distillation Adapters
This distillation adapter is for use with bench or pilot plant reactors.
  • Center vessel collects moisture
  • With bottom PTFE plug stopcock
  • Fabricated from borosilicate glass
Moisture is collected in the center vessel and drained off through the bottom stopcock which is ground to accept a compression style fitting. Stopcock is a PTFE plug. Available with either one or two reinforced top standard taper outer joints, and single standard taper inner joint at bottom next to stopcock. Joint Size value represents the compression fitting joint size .
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty