Would you like to log in again?
Duran Bottle GL 45 Connection System, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7632-06 7632-12 7632-05 7632-01 7632-10 7632-07 7632-11 7632-02 7632-13
89214-902PK 19.63 USD InStock
89214-902 89214-936 89214-914 89214-900 89214-944 89214-912 89214-904 89214-940 89214-930
Duran Bottle GL 45 Connection System, Ace Glass Incorporated
Closures Bottle Closures
The heart of the GL45 connection system is a GL45 polypropylene cap with GL18 threaded ports
Modular accessories include polypropylene tube connection caps,solid polybutylene terepthalate (PBT) caps, PBT pressure equalization caps and PTFE cap inserts to accomodate various sizes of tubing. A typical application is the safe transfer of liquid media within a closed and sterile system for which you'd need one two-pack GL45 cap, two 89214-900 equalization caps, and some combination of solid caps and tubing connections with inserts.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty