Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Flask, Single Neck, Ace Glass Incorporated
Supplier: Ace Glass
6887-31 6887-241 6887-21 6887-37 9458-214 6887-27 6887-230 9458-212 6887-225 9458-21 9458-23 9458-12 9458-204 6887-233 9458-223 9458-08 6887-242 6887-220 6887-235 6887-244 6887-237 6887-72 9458-09 9458-221 6887-23 6887-228 6887-65 6887-263 6887-29 9458-206 6887-20 6887-40 6887-221 9458-17 6887-53 6887-41 9458-202 6887-28 6887-253 6887-203 6887-236 6887-66 6887-234 6887-238 9458-208 6887-24 6887-223 6887-262 6887-68 9458-210 6887-38 9456-06 6887-231 9458-05 9458-215 9458-217 9456-03 6887-35 9458-06 6887-232 6887-207 9458-225 6887-264 6887-64 6887-42 6887-245 9458-07 6887-70 9458-19 6887-265 6887-227 9458-219 9458-25 6887-44 9458-10 6887-69 9458-04 6887-30 6887-63 6887-25 6887-226 6887-67 6887-26 6887-43 9458-15 9458-14 9458-205 9458-02 6887-243 6887-239 6887-240 6887-36 6887-224 6887-82 6887-229
89074-586EA 48.02 USD InStock
89074-586 89074-620 89074-584 89074-624 89074-588 89074-622 89083-620 89074-628 89056-180 89056-260 89056-182 89074-626 89056-266 89056-222 89056-188 89056-268 89056-224 89056-262 89056-184 89056-220 89056-264 89056-186 89074-582 89056-226 89056-228 89074-580 89074-630 89074-598 89074-596 89074-634 89074-632 89056-192 89074-638 89215-088 89074-636 89056-194 89083-610 89056-270 89083-612 89056-190 89083-614 89056-276 89056-232 89056-234 89056-278 89056-196 89056-272 89056-198 89056-230 89056-274 89074-590 89074-594 89056-236 89056-238 89074-592 89074-642 89083-608 89074-640 89074-602 89074-600 89056-280 89074-606 89056-282 89074-604 89074-608 89056-244 89056-288 89056-200 89056-202 89056-246 89056-284 89056-240 89056-242 89056-286 89056-208 89056-204 89056-248 89056-206 89074-612 89074-610 89074-616 89074-614 89074-618 89056-178 89056-210 89056-254 89056-212 89056-256 89056-250 89056-176 89056-252 89056-218 89056-258 89056-214 89056-216
Flask, Single Neck, Ace Glass Incorporated
Flasks Round Bottom Flasks
These round bottom flasks have a single short neck with a [ST] outer joint
Some sizes are available with heavy walls, approximately 30% heavier than standard-wall flasks. Full-length standard taper joints (only) are reinforced for better strength. Fabricated from borosilicate glass.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty