Would you like to log in again?
Photochemical Jacketed Reactor with Ace-Thred (No Valve), Ace Glass Incorporated
Supplier: Ace Glass
7864-08 7864-10 7864-12
89215-144EA 1670.79 USD InStock
89215-144 89215-146 89215-148
Photochemical Jacketed Reactor with Ace-Thred (No Valve), Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Jacketed reaction vessel designed to enable cooling of reactant material during photolysis
Constructed of borosilicate glass to accommodate the immersion wells. With one standard taper 14/20 angled joint for sparger tube, one standard taper 24/40 vertical joint for condenser and one #7-Ace-Thred side arm for thermometer. Center joint is #50 Ace-Thred. Bottom of reactor is flat to allow use of stir bar. Volumes indicated are total volumes. Volume in reactive area of lamp is approximately 40-50% of total volume. Size C hose connections. Listing is vessel only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty