Would you like to log in again?
Photochemical Jacketed Reactor with Ace-Thred and Stopcock Bottom, Ace Glass Incorporated
Supplier: Ace Glass
7865-06 7865-10 7865-08
89215-150EA 2103.25 USD InStock
89215-150 89215-152 89215-154
Photochemical Jacketed Reactor with Ace-Thred and Stopcock Bottom, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Jacketed reaction vessel to enable cooling of reactant material during photolysis plus the added feature of a 2 mm bore, 1:5 PTFE stopcock at bottom for draining inner vessel contents
Constructed of borosilicate glass to accommodate the immersion wells. With one- standard taper 14/20 angled joint for sparger tube, one- standard taper 24/40 vertical joint for condenser and one-#7 Ace-Thred side arm for thermometer. Center joint is #50 Ace-Thred. Bottom of reactor is flat to allow use of stir bar. Volumes indicated are total volumes. Volume in reactive area of lamp is approximately 40- 50% of total volume. Size C hose connections. Listing is vessel only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty