Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Test Tube Racks
Supplier: VWR
202112-G 202115-B 201013-B 201010-G 202112-O 202114-R 202114-Y 202111-W 201012-G 202110-W 202110-R 202113-G 201011-R 201010-R 202113-W 202112-B 202111-B 202116-B 201012-W 201014-W 202113-O 202113-B 202114-G 202116-G 202116-R 202112-R 201010-B 202111-G 202110-B 201010-W 202111-O 202115-W 202113-Y 201011-W 201012-R 202115-G 201014-R 202111-R 202112-Y 202112-W 202110-Y 201012-B 202114-W 201014-B 201011-B 202113-R 202116-W 202110-M 202115-M 201013-W 201011-G 202110-G 201014-G 202114-O 202110-O 202111-Y 202114-B
89215-770CS 234.43 USD InStock
89215-770 89215-772 89215-774 89215-730 89215-732 89215-776 89215-810 89215-800 89215-768 89215-724 89215-802 89215-726 89215-804 89215-728 89215-806 89215-808 89215-760 89215-762 89215-720 89215-764 89215-766 89215-722 89215-756 89215-758 89215-716 89215-718 89215-790 89215-792 89215-750 89215-794 89215-752 89215-796 89215-798 89215-754 89215-822 89215-746 89215-824 89215-748 89215-826 89215-828 89215-780 89215-782 89215-740 89215-784 89215-786 89215-742 89215-820 89215-788 89215-744 89215-734 89215-778 89215-812 89215-736 89215-814 89215-738 89215-816 89215-818
VWR® Test Tube Racks
Racks Tube Racks
VWR® Test Tube Racks are available in both nylon glass filled or polypropylene options.
  • Both glass filled or polypropylene options are easy to handle
  • Racks may be used in an incubator, fume hood, refrigerator, or for benchtop applications
  • Autoclavable
  • No assembly required
Racks are a single piece molded from either nylon 6 glass fiber reinforced material, or polypropylene.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty