Would you like to log in again?
AirWave® 4200 Series N95 Particulate Respirators, Moldex®
Supplier: Moldex
AirWave®
4200
89216-291CS 382.58 USD InStock
89216-291
AirWave® 4200 Series N95 Particulate Respirators, Moldex®
Respirators Disposable Respirators Particulate Respirators
Easier breathing and cool comfort are what you get when you wear an AirWave® respirator
  • Unique FlexFit® design moves with the mouth for comfort all day long
  • Pinch-free molded nose bridge eliminates metal nose-band readjustment and discomfort
  • Dura-Mesh® shell prevents collapsing from heat and humidity so the mask lasts longer
The larger surface area of the patent-pending wave design means more air flows in and out of the mask - all without a valve! Easy breathing… by design.
Certifications: NIOSH certified.
Ordering information: Respirators are packaged 10 per box, 8 boxes per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty