Would you like to log in again?
VWR® Amber HDPE Carboys with Spigots
Supplier: VWR
15K-5221-VWR 15K-1211-VWR 15K-3221-VWR 15K-2211-VWR
89217-234EA 521.55 USD InStock
89217-234 89217-236 89217-230 89217-232
VWR® Amber HDPE Carboys with Spigots
Bottles Carboys
VWR® amber carboys meet USP standards for light-transmission, making them ideal for storing photo-sensitive materials, while providing users with a highly versatile container that maximizes storage efficiency and ease of use.
  • Conserve benchtop space with rectangular shape
The distinctive rectangular shape saves valuable bench space.
VWR® amber carboys are available in HDPE, offering excellent chemical resistance. VWR® amber carboys are available with or without a spigot. Models with a spigot are compatible with versatile Barbs, which create bottom tubulation options in a variety of hose barb sizes. Versatile caps, adapters, versatile Barbs, and spigots are constructed of polypropylene and are autoclavable at 15 psi. and 121 °C (250 °F) for 20 minutes.
Large handles on top of the container and molded handles in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional 120 mm (4³/₄") wide neck allows users to reach inside the carboy for easier cleaning. Leakproof polypropylene versatile caps offer the ultimate in connection flexibility. Open caps (sold separately) are compatible with interchangeable adapters, which feature a wide variety of top-connections, including molded-in hose barbs and quick-connect hose barbs. Carboy material and volume are clearly imprinted on the outside of the bottle as well as large, easy to read, metric graduation marks which are certified to ±5% accuracy.
Certifications: Compliance with FDA requirements for food use.
Ordering information: Carboys are supplied with a closed cap; open caps are sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty