Would you like to log in again?
SP Bel-Art Break-Safe™ Ampule Opener, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
Scienceware®
F389520000
89217-378CS 925.93 USD InStock
89217-378
SP Bel-Art Break-Safe™ Ampule Opener, Bel-Art Products, a part of SP
Ampule Openers
Safely open up to three ampules at once with the Break-Safe™ Ampule Opener.
  • Cleanly snapping ampule necks
  • Copolyester plastic construction
  • Broad chemical resistance
  • Unobstructed viewing of ampule contents
Unique box-style design allows one-handed opening of up to 3 ampules at a time by simply pressing down on lid. Ampule necks snap clean and are contained within the device while fingers stay away from sharp edges. After removing ampule contents, turn the device upside down over a trash receptacle to safely discard ampules and tops.
Ordering information: Includes 4 inserts to allow opening of 1, 2, 5 and 10 ml ampules.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty