Would you like to log in again?
Prefilled Bouin's Solution, Azer Scientific
Supplier: Azer Scientific
PFB-020 PFB-040 PFB-060
89217-510CS 278.38 USD InStock
89217-510 89217-513 89217-515
Prefilled Bouin's Solution, Azer Scientific
Cell Culture Supplements and Additives
Bouin's fixative is a saturated picric acid-formalin solution that is useful for preserving and fixing delicate tissues and gastrointestinal tract specimens.
  • Available in 20, 40 and 60 ml containers each half-filled
  • 95 kPa compliant specimen containers
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty