Would you like to log in again?
Bouin's Solution, Azer Scientific
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B-F-G CUNB-5G CUNB-2.5G
89217-520EA 400.21 USD InStock
89217-520 89217-516 89217-518
Bouin's Solution, Azer Scientific
Stains Microscopy and staining reagents
Bouin's solution is a saturated picric acid-formalin fixative that is useful for preserving delicate tissues and gastrointestinal tract specimens.
Packaging: Dispending spigots are included with cube containers.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty