Would you like to log in again?
Measuring Cylinders, To Contain, Class A, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20027-2000 20027-1000 20027-50 20027-10 20027-500 20027-25 20027-100 20027-250
89217-674CS 507.48 USD InStock
89217-674 89217-664 89217-676 89217-670 89217-672 89217-666 89217-678 89217-668
Measuring Cylinders, To Contain, Class A, Kimble Chase
Cylinders Measuring Cylinders
Class A, to contain, measuring cylinders feature a pour spot top
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: A SAFE-GARD® bumper is supplied with each measuring cylinder.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty