Would you like to log in again?
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Heavy Duty, Wide Mouth, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
92820G-25 92820G-20 92820G-250 92820G-500 92820G-1000 92820G-10 92820G-200 92820G-50 92820G-5 92820G-2000 92820G-100
89217-684CS 418.53 USD InStock
89217-684 89217-696 89217-686 89217-698 89217-680 89217-692 89217-682 89217-694 89217-690 89217-688 89217-700
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Heavy Duty, Wide Mouth, Kimble Chase
Flasks Volumetric Flasks
These KIMAX® flasks are permanently marked with an individual serial number and are supplied with a Certificate of Graduation Accuracy
Designed from ASTM Specification E288, Class A serialized requirements. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Pipet access is easy with these wide-mouth volumetric flasks. Heavy, uniform walls provide superior strength, durability and safety. Large, permanent, easy-to-read markings allow for quick identification. Graduated to Class A volumetric tolerances with graduation ring blasted on the neck. Calibrated "to contain." Flasks feature flat interior bottoms for stirring with standard stir bars. Supplied with a marking spot and standard taper glass stopper.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty