Would you like to log in again?
Virtua™ Protective Eyewear, 3M™
Supplier: 3M
Virtua™
11331-00000-20 7100112434 7100114652 7010386340 7100123356 7100109223
47746-241CS 88.74 USD InStock
47746-241 47746-233 89217-718 47746-237 89107-432 89006-547
Virtua™ Protective Eyewear, 3M™
Safety Glasses
Virtua™ Protective Eyewear provides unisex styling, protection, and affordability for site visitors or single-use protection.
  • Designed for indoor or outdoor use
  • Wide profile provides necessary impact protection and an unobstructed view
Lightweight, unisex Virtua™ Protective Eyewear combines versatility and value with high wraparound coverage, polycarbonate construction, and a hard coat, scratch-resistant lens for a comfortable, contoured fit.
Certifications: ANSI Z87.1.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty