Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Bel-Art FRIO-Temp® High Precision Verification Thermometer, Bel-Art Products, a part of SP

Supplier: Bel-Art Products, a part of SP

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

20400 20402 20404 20401 20403
89217-860EA 123.24 USD InStock
89217-860 89217-862 89217-858 89217-864 89217-856
SP Bel-Art FRIO-Temp® High Precision Verification Thermometer, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
FRIO-Temp® Thermometers are used to verify temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST.

  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Insert thermometer into the bottle and place near the samples for best results
  • Feature permanently fused markings and white lead free glass

Each item features PFA safety coating, a multi-language (English, French, German, Italian, Portuguese, Spanish) Statement of Accuracy and Traceability and individual serial number. Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Certifications: NIST.

Order Now
 

ORDER

SERVICES


Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to our certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty