Would you like to log in again?
SP Bel-Art Frio-Temp® Calibrated Electronic Verification Thermometers for Freezers, Refrigerators, Incubators and Ovens, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FRIO-Temp®
B60210-0800 B60210-0900 B60210-0500 30715 B60210-0400 30700 B60210-0700 B60210-1000 B61000-0600 B60210-1100 30716 B61000-5000 B60210-0600 B60210-0300
10753-244CS 699.39 USD InStock
10753-244 89217-956 75854-360 75854-370 75854-362 75854-364 75854-352 75854-366 75854-354 75854-368 75854-356 89217-960 89217-962 75854-358
SP Bel-Art Frio-Temp® Calibrated Electronic Verification Thermometers for Freezers, Refrigerators, Incubators and Ovens, Bel-Art Products, a part of SP
Thermometers Digital Thermometers
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)
  • Individually serialized, CE marked
Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Caution: Note: Plastic bottles as indicated in table cannot be used in ovens.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty