FRIO-Temp® Thermometers are used to verify temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST, DKD/PTB and most standards through ILAC.
- White back glass
- Organic thermometer liquid fill
- Highly accurate readings
- PFA safety coating
Each item features PFA safety coating, a detailed Calibration Report stating corrections to one-tenth of the smallest division and individual serial number. Each item is tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST and DKD/PTB. Individually packaged. Glass Color: White. Organic thermometer fill. Enviro-Safe® liquid bottle fill.
Certifications: NIST, DKD/PTB