Would you like to log in again?
SP Bel-Art FRIO-Temp® Certified Precision Liquid-In-Glass Verification Thermometer, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20715C 20630C 20700C 20720C 20717C
89217-994EA 374.62 USD InStock
89217-994 89217-984 89217-990 89217-986 89217-998
SP Bel-Art FRIO-Temp® Certified Precision Liquid-In-Glass Verification Thermometer, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
FRIO-Temp® Thermometers are used to verify temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST, DKD/PTB and most standards through ILAC.
  • White back glass
  • Organic thermometer liquid
  • Highly accurate readings
  • PFA safety coating
  • Individually packaged
Each item features PFA safety coating, a detailed Calibration Report stating corrections to one-tenth of the smallest division and individual serial number. Each item is tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST and DKD/PTB. Individually packaged. Glass Color: White.
Certifications: NIST, DKD/PTB
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to our certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty