FRIO-Temp® Thermometers are used to verify temperatures in freezers, refrigerators, incubators and ovens with traceability to most ILAC standards.
- Each item features PFA safety coating
- Each item is tested in H-B's exclusive triple accredited/registered ISO/IEC 17025:2017 A2LA accredited laboratory
- Individually packaged
A2LA accredited laboratory against equipment whose calibration is traceable to certifications.
Certifications: Thermometers are traceable to NIST.
Delivery information: A multi-language (English, French, German, Italian, Portuguese, Spanish) Statement of Accuracy available.