Would you like to log in again?
SP Bel-Art Double-Safe PFA Safety Coated, Environmentally Friendly Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
30599T 30545T 30592T 30591T 30535T 30596T 30515T 30510T 30505T 30593T 30540T
89218-032EA 49.78 USD InStock
89218-032 89218-022 89218-042 89218-036 89218-026 89218-034 89218-046 89218-038 89218-016 89218-050 89218-040
SP Bel-Art Double-Safe PFA Safety Coated, Environmentally Friendly Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
These environmentally safe thermometers are PFA safety coated to keep liquid and glass contained in the event of breakage.
  • Organic liquid fill
  • Non-toxic and biodegradable
  • Lead-free
  • PFA safety coating
  • Yellow back glass
Non-toxic, biodegradable, Enviro-Safe® liquid, against lead-free, yellow back glass provides easy readability and fewer reading errors. They are ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Ordering information: Manufactured and tested by our USA quality team, these thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability. Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to our certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty