Would you like to log in again?
Glove Box, Closed-Loop Containment, PLAS-LABS™
Supplier: Plas-Labs
830-ABB/CLC 830-ABD/CLC 830-ABC/CLC
89218-120EA 17858.39 USD InStock
89218-120 89218-124 89218-122
Glove Box, Closed-Loop Containment, PLAS-LABS™
Glove Boxes and Glove Bags
This Closed-Loop Containment glove box minimizes drafts and maximizes effectiveness of your balance
Product Benefits: Removable top section, four purging gas valves, white ambidextrous gloves, hospital grade outlet strip, transfer chamber 12" long x 11" ID.
Basic features: Two (2) 8" round HEPA filters, blower, outlet strip, Magnehelic gauges.
This unit features cleanliness, safety, complete containment, portability, and a draft-free atmosphere.The Compact Glove Box is designed specifically for use in hazardous situations where materials to be weighed must be contained.
Certifications: UL Listed.
Ordering information: Glove box comes ready to use, no installation required. Accessory kit includes plug and case.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty