Would you like to log in again?
Cryo.s™ Storage Box, Rack and Lid, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Cryo.s™
802225 802501 802206 802204 802203 802202
89218-652CS 150 USD InStock
89218-652 82051-490 89218-650 89176-318 89218-654 89218-648
Cryo.s™ Storage Box, Rack and Lid, Greiner Bio-One
Boxes Cryoboxes
Greiner Bio-One offers the Cryo.s™ Storage Box is a helpful accessory for storing Cryo.s™ sample tubes at low and ultra-low temperatures.
  • Polypropylene construction
  • Shock-resistant
  • Resilient to extremely low temperatures
The box material polypropylene is very temperature and shock-resistant, thus allowing storage temperatures as low as -196 °C.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty