Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Bel-Art Earth-Friendly '1001' Sampling Sticks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SCIENCEWARE®
F36931-0000 F36931-0001
89218-784CS 292.5 USD InStock
89218-784 89218-782
SP Bel-Art Earth-Friendly '1001' Sampling Sticks, Bel-Art Products, a part of SP
Spatulas
These earth-friendly sampling sticks are durably constructed with 100% PLA (polylactic acid) resin.
  • PLA resin construction
  • Earth-friendly with no petroleum-based materials
  • Equally as strong as non-biodegradable plastic resins
PLA resin is a plant-based material and is a renewable resource with little environmental impact. PLA offers a lower carbon footprint, and is 100% biodegradable through industrial composting. Sampling sticks made from PLA polymers are just as strong as sampling sticks made from other non-biodegradable plastic resins. These earth-friendly sticks contain no petroleum-based materials and contribute far less greenhouse gases.
14.6L x 1.9W x 0.15H cm (5¾ x ¾ x ¹⁄₁₆")
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty