Would you like to log in again?
Permafrost Adhesion Economy Frosted Microscope Slides, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
940BO-ADH-001 940T-ADH-001 640-ADH-001 646-ADH-001 643-ADH-001 642-ADH-001 644-ADH-001
89260-794CS 743.51 USD InStock
89260-794 89218-838 89260-800 89218-840 89218-844 89218-842 89218-836
Permafrost Adhesion Economy Frosted Microscope Slides, Thermo Scientific
Slides Adhesive Slides
Permafrost Adhesion Slides are designed for tissue adhesion in routine clinical applications
All Permafrost Adhesion slides are pre-cleaned, measure 25x75x1mm, feature smoothly ground edges, and are manufactured from white Swiss glass. Slides are packed approximately 72 per pack, 20 packs per case.
Permafrost Adhesion slides feature a ¾” frosted end with two "a" markings at the bottom of each slide. They are available in nine different colors.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty