Would you like to log in again?
NorthFlex™ Light Task Plus II™ Polyurethane Palm Coated Dyneema® Gloves, Honeywell Safety
Supplier: Honeywell Safety
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NFD15B/7S NFD15B/8M NFD15B/6XS NFD15B/10XL NFD15B/11XXL NFD15B/9L
89219-010PK 256.44 USD InStock
89219-010 89219-012 89219-014 89219-016 89219-018 89219-008
NorthFlex™ Light Task Plus II™ Polyurethane Palm Coated Dyneema® Gloves, Honeywell Safety
Gloves
Gloves feature a Dyneema® seamless liner with white polyurethane coating and knit wrists.
  • Dynemma® cut-resistant fiber shell
  • White with white polyurethane palm and fingertip coating
  • Gloves provide a good cut and abrasion protection level
Lightweight gloves are ideally suited for use in construction, manufacturing, municipal services, and steel and metals applications and industries.
Certifications: Meets ANSI/ISEA 105 - ASTM D3389-05, ANSI/ISEA 105 - ASTM F1790-97, EN 388, and EN 420 requirements.
Packaging: Gloves are packaged 12 pair per pack, 72 pair per case.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty