Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Reli® PGA Violet Braided Absorbable Sutures, MYCO Medical
Supplier: MYCO Medical
V466 V442
89219-076BX 218.55 USD InStock
89219-076 89219-080
Reli® PGA Violet Braided Absorbable Sutures, MYCO Medical
Sutures
Reli® PGA Violet Braided Sutures feature high quality, high performance needles made from AISI 304 Stainless Steel for improved needle integrity and sharpness, smooth insertion, and patient comfort
These sutures also exhibit high knot security and tensile strength. A full range of sutures is available to meet physician preference.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty