Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Reli® Black Nylon Non-Absorbable Sutures, MYCO Medical
Supplier: MYCO Medical
N1665 N928 N6681 N667 N697 N1663 N1666 N662 N661 N6621 N664 N699 N663
89219-114BX 183.16 USD InStock
89219-114 89219-126 89219-104 89219-106 89219-118 89219-108 89219-130 89219-120 89219-098 89219-110 89219-100 89219-134 89219-102
Reli® Black Nylon Non-Absorbable Sutures, MYCO Medical
Sutures
Reli® Black Nylon Sutures feature high quality, high performance needles made from AISI 304 Stainless Steel for improved needle integrity and sharpness, smooth insertion, and patient comfort
These sutures also exhibit high knot security and tensile strength. A full range of sutures is available to meet physician preference.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty