Would you like to log in again?
Caps for Mobile Phase Bottles, Microsolv
Supplier: MicroSolv
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
58807-B45 58807-G45 58807-P45 58807-Y45
89219-970PK 57.49 USD InStock
89219-970 89219-972 89219-974 89219-976
Caps for Mobile Phase Bottles, Microsolv
Closures Bottle Closures
These caps are designed for use on HPLC mobile phase reservoir and storage bottles with GL 45 thread.
  • Made up of polypropylene
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty