Would you like to log in again?
SEALING RING F/MP BOTTLEPK100
Supplier: MicroSolv Technology Corp
Rings, drip free^ Sealing for HPLC mobile phase reservoir and storage bottles with GL 32 threads. Polypropylene. 10/pack.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SEALING RING F/MP BOTTLEPK100 89219-990 58807-Z32 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty