Would you like to log in again?
CAP SCREW TOP PRESLIT SIN/PTFE YL PK100
Supplier: MicroSolv Technology Corp
Caps, screw with Preslit silicone/PTFE septa^ Fitted in smooth cap. Yellow, for use with 9-425mm, 12x32mm autosampler vials. 100/pk
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP SCREW TOP PRESLIT SIN/PTFE YL PK100 89220-004 9502S-35-Y Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty