Would you like to log in again?
VWR® Vacuum Filtration Systems featuring Pall Membranes
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
146-1223-VWR 1151-VWR 1162-VWR 116-9123-VWR 1101-VWR 1122-VWR 116-1123-VWR 146-2223-VWR 116-3123-VWR 1412-VWR 146-1123-VWR 1141-VWR 1411-VWR 1402-VWR 1153-VWR 1143-VWR 1103-VWR 1102-VWR 146-4123-VWR 116-2123-VWR 116-8123-VWR 146-2323-VWR 1403-VWR 146-5123-VWR 1116-VWR 1161-VWR 146-2123-VWR 1152-VWR 1142-VWR 1163-VWR 116-7123-VWR
89220-716CS 405.77 USD InStock
89220-716 97066-206 89220-714 97066-208 89220-712 97066-202 89220-710 97066-204 97066-220 89220-698 97066-200 89220-696 97066-222 89220-694 89220-708 97066-216 89220-728 97066-218 89220-704 89220-726 97066-212 97066-234 89220-702 89220-724 97066-214 89220-700 89220-722 89220-720 97066-210 97066-232 89220-718
VWR® Vacuum Filtration Systems featuring Pall Membranes
Filters Bottle Top Filters
VWR® Vacuum Filtration Systems are designed for the preparation of buffers, tissue culture media, microbiological media, and other biological fluids.
  • For the preparation of biological fluids
  • Feature Pall membranes
  • Compatible with the quick-connect vacuum pedestal for use with existing laboratory stands
They feature large diameter Pall membranes for guaranteed performance and maximum throughput.
The centrifuge tube filtration system includes a filter funnel, 15 or 50 ml centrifuge tube with polypropylene screw cap, and vacuum hose adapter. The system is designed to allow the user to filter fluids directly into the tube, eliminating the potential for contamination and spills during post-sterilization transfer. The bottle-top filtration system includes a receiver bottle with an ergonomic shape, molded finger grips, and a patented bottle cap.
All systems are compatible with the quick-connect vacuum pedestal, vacuum adapter (included), and cradle ring for use with existing laboratory stands. All three options allow for a convenient one-time vacuum hose connection, and one-handed operation. All items are available with 0.1 µm (Mycoplasma filtering grade), 0.2 µm (sterilizing grade) or 0.45 µm (clarification grade) asymmetric PES membranes. All items are made from Class VI materials, and are conveniently sold gamma sterilized.
Ordering information: Vacuum filtration pedestal and vacuum filtration cradle ring are sold separately.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty