Would you like to log in again?
Chemically Inert HV Standard Manual Valves, Hamilton Company
Supplier: Hamilton
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
86788 86779 86913 86735 86787 86786 86916 86729 86912 86738 86904 86780 86732 86908 86778 86785 86915 86918 86905 86776 86910 86903 86901 86911 86725 86726 86731 86907 86736 86728 86781 86737 86775 86727 86730 86777
65512-700EA 274.02 USD InStock
65512-700 65510-636 89221-328 89221-324 89221-368 89221-346 89221-348 89221-326 89221-364 89221-342 89221-320 89221-344 89221-366 89221-322 89221-360 89221-362 89221-340 65511-195 65510-953 89221-318 65510-159 65510-115 89221-336 89221-358 65510-035 65510-352 89221-338 65510-670 89221-354 89221-332 65510-272 89221-356 89221-334 65510-590 89221-350 89221-330
Chemically Inert HV Standard Manual Valves, Hamilton Company
Valves Vacuum Valves
Hamilton Company offers the most comprehensive selection of standard and custom syringes, valves and accessories in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. Hamilton (HV) Plug Valves provide an economical and flexible method of custom-building a fluid flow control system, from a simple on/off valve to a more complex multi-valve arrangement, using a combination of loop and distribution valves. The small valve body with its chemically inert flow path is ideally suited for low pressure applications. All HV Plug Valves allow you to select and interchange a wide variety of fittings, adapters and tubing, including male and female luers, FEP tubing assemblies, and adapters which thread directly into the valve providing an endless variety of valve configurations for any flow application. Hamilton (HV) fittings and adapters are installed finger-tight (10 ounce inches) and provide a leak-tight seal tested to 100 psig.
The HV valve is physically the smallest valve and constitutes a basic unit for the control of fluids. It is ideal for low pressure applications; from a basic on/off flow to a four-way distribution system. They have a threaded path of 1/4-28. Their operator pressure is 0 to 100 psig (0 to 6.9 bar), and their fluid path is PTFE/CTFE. The HVP valve is essentially the same as the HV valve except the HVP can be mounted into panels allowing for custom installation into control panels or instruments.
HVX valves are physically the largest valves. A larger port internal diameter allows increased fluid flows even with moderately viscous fluids. HVX valves can be used free standing or panel-mounted using an optional locking nut. 6- and 8-port distribution and loop valves are available only in the HVX body style.
Instrument configurations include HV - Hamilton Valve, Small Body; HVP - Hamilton Valve, Panel Mount, Small Body; HVX - Hamilton Valve Standard or Panel Mount, Large Body;
Flow valves are used to direct or redirect fluids. 2-, 3-, and 4 -port valves are available with straight through (180°), 90°, or "T" type plugs for seven different valves. All seven flow configurations are available as HV, HVP and HVX valves. The flow configuration drawings show the possible fluid flow paths for a three port flow valve with a "T" type plug. Loop valves are used to direct and redirect fluids. 4-, 6-, or 8- port valves are available with a loop type valve plug. The 4- port valve is available in HV, HVP and HVX body styles. The 6- and 8-port valves are only available in the HVX body style. The flow configuration drawings show the possible fluid flow paths for a 4-port valve with a loop type plug. Distribution valves are used to direct fluid to a number of locations. 2-, 3-, 4-, 6- and 8-port valves are available with a distribution type valve plug. The 2-, 3-, and 4-port distribution valves are available in HV, HVP and HVX body styles. The 6- and 8-port valves are available only in the HVX body style. The flow configuration drawings show the possible fluid flow paths for a 4-port valve with a distribution type plug.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty