Would you like to log in again?
Syringe Cleaner, Hamilton
Supplier: Hamilton
76615
89221-464EA 388.24 USD InStock
89221-464
Syringe Cleaner, Hamilton
Cleaning Agents Detergents and Surface Cleansers
Specifically designed for rapidly cleaning 7000 series modified Microliter™ syringes.
Generally, Microliter™ or Gastight® syringes require a lower cleaning temperature than the Modified Microliter™ syringes
This syringe cleaner also effectively cleans Microliter™ and Gastight® syringes. A combination of heat (approximately 370 °C) and vacuum vaporises and extracts contaminants from the needle.
Note: Do not heat the syringes with cemented needles above 50 °C. Thermal expansion of the needle will fracture the glass. Do not allow the glass barrel to come into contact with the hot septum cap.
It is ineffective for septa-plugged needles, seized plungers and other irreparable needles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty