Would you like to log in again?
Sterility Testing Medium-A, HiMedia Laboratories
Supplier: HiMedia Laboratories
M017-500G M017-500G M017-5KG M017-2.5KG M017-2.5KG M017-5KG
95020-050EA 92.26 USD InStock
CA95020-050 95020-050 CA89222-218 89222-218 CA89222-220 89222-220
Sterility Testing Medium-A, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
This sterility testing medium A is used for checking presence of aerobic microorganisms in pharmacopoeial articles.
The medium contains peptic digest of animal tissue and meat extract which provide carbonaceous, nitrogenous compounds and essential growth nutrients for the growth of microorganisms. Sodium chloride maintains the osmotic balance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty