Would you like to log in again?
XLD Agar, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
MH031-2.5KG MH031-2.5KG MH031-5KG M031-500G MH031-5KG
89222-274EA 808.09 USD InStock
89222-274 CA89222-276 CA89222-274 61000-000 89222-276
XLD Agar, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Xylose-Lysine Deoxycholate Agar (XLD Agar) is a selective medium recommended for the isolation and enumeration of Salmonella Typhi and other Salmonella species from clinical and non clinical samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty