Would you like to log in again?
Koser Citrate Medium, HiMedia Laboratories
Supplier: HiMedia Laboratories
M069-500G M069-500G M069-2.5KG M069-5KG M069-2.5KG M069-5KG
89222-402EA 1112.81 USD InStock
89222-402 CA89222-400 89222-400 CA89222-402 CA61000-002 61000-002
Koser Citrate Medium, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Koser citrate medium is used to differentiate Escherichia coli from Enterobacter aerogenes on the basis of citrate utilization.
  • White to cream colored
  • homogeneous, free flowing powder
  • Reaction of 0.57 w/v aqueous solution at 25 °C
Caution: Store below 30 °C in tightly closed container and prepared medium at 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty