Would you like to log in again?
Nutrient Agar 1.5%, HiMedia
Supplier: HiMedia Laboratories
M087-5KG M087-5KG M087-500G M087-2.5KG M087-500G M087-2.5KG
89222-502EA 521.85 USD InStock
89222-502 CA89222-504 89222-504 CA89222-502 CA95020-228 95020-228
Nutrient Agar 1.5%, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Nutrient Agar is recommended for cultivation of bacteria not requiring a highly nutritious medium can be enriched with blood, ascitic fluid or other enriching fluids.
  • Final pH ( at 25 °C) is 7.3 ± 0.2
  • Cream to yellow homogeneous free flowing powder
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty