Would you like to log in again?
Nutrient Agar, 1.5%, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M087I-5KG M087I-2.5KG M087I-500G M087I-2.5KG M087I-5KG M087I-500G
89222-506EA 573.95 USD InStock
89222-506 CA95020-230 95020-230 CA89222-508 CA89222-506 89222-508
Nutrient Agar, 1.5%, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Recommended as a general-purpose nutrient medium which can be used for cultivation of fastidious microorganisms after appropriate enrichment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty