Would you like to log in again?
Lactose Broth, Himedia
Supplier: HiMedia Laboratories
M1003S-2.5KG M1003S-2.5KG
89222-574EA 552.06 USD InStock
CA89222-574 89222-574
Lactose Broth, Himedia
Media for Microbiology Dehydrated Media for Microbiology
Lactose Broth is used for the detection of coliform bacteria in water, foods, and dairy products as per Standard Methods.
  • Recommended by APHA in the performance and confirmation of the presumptive test for coliform bacteria in water, food and milk
  • Provides excellent results in Eijkman Assays of gas production
  • In testing dairy products, Lactose Broth is used only in the completed test
Lactose Broth is recommended by APHA in the performance and confirmation of the presumptive test for coliform bacteria in water, food and milk. This medium was initially listed as an alternative to Lauryl Sulfate Broth in the presumptive Standard Total Coliform Multiple-Tube (MPN) Test for water analysis. Although it is not the original formulation, Lactose Broth provides excellent results in Eijkman Assays of gas production at 45° C, which is a characteristic of Escherichia coli. While preparing this medium it is important to avoid overheating and to distribute it into tubes before sterilization. Peptic digest of animal tissue and beef extract in the medium supply essential nutrients to the organisms. Lactose is a fermentable carbohydrate for the coliforms. Tubes of Lactose Broth are inoculated with dilutions of water or milk, etc. under test, and incubated at 35° C and examined for gas formation after 24 and 48 hours. Members of the coliform group are defined as aerobic and facultative anaerobic gram-negative and non-sporing bacilli, which ferment lactose with gas formation within 48 hours at 35° C. In testing dairy products, Lactose Broth is used only in the completed test. Large water samples may require double strength Lactose Broth to minimize the final volume.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty