Would you like to log in again?
Dey-Engley Neutralizing Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1062-5KG M1062-2.5KG M1062-500G M1062-500G M1062-2.5KG M1062-5KG
95020-396EA 278.08 USD InStock
CA89222-784 CA95020-396 95020-396 89222-784 CA89222-782 89222-782
Dey-Engley Neutralizing Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Dey-Engley Neutralizing Broth is used in disinfectant testing where neutralization of the antiseptics and disinfectants is important for determining its bactericidal activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty