Would you like to log in again?
Tryptone Yeast Extract Agar, HiMedia
Supplier: HiMedia Laboratories
M1193-5KG M1193-500G M1193-500G M1193-2.5KG M1193-5KG M1193-2.5KG
89223-282EA 918.21 USD InStock
89223-282 89223-284 CA89223-282 CA89223-284 CA95020-668 95020-668
Tryptone Yeast Extract Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Tryptone yeast extract agar with BCP is used for isolation and enumeration of Enterobacteriaceae and Bacillus cereus.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty