Would you like to log in again?
Nutrient Agar No.2, Himedia
Supplier: HiMedia Laboratories
M1269-5KG M1269-500G M1269-5KG M1269-2.5KG M1269-500G M1269-2.5KG
89223-554EA 574.18 USD InStock
89223-554 89223-556 CA89223-554 CA89223-556 95020-830 CA95020-830
Nutrient Agar No.2, Himedia
Media for Microbiology Dehydrated Media for Microbiology
Nutrient media are general purpose media used for the examination of water and dairy products.
  • Cream to yellow homogeneous free flowing powder
  • Firm, comparable with 1.5% agar gel
  • Maintenance of subcultures
Nutrient media are general purpose media used for the examination of water and dairy products. It can also be used for sterility testing of aerobes and also for maintenance of subcultures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty