Would you like to log in again?
Synthetic Sea Salt, HiMedia Laboratories
Supplier: HiMedia Laboratories
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M1344-2.5KG M1344-5KG M1344-500G M1344-500G M1344-2.5KG M1344-5KG
89223-780EA 783.27 USD InStock
CA89223-782 CA95020-976 89223-780 CA89223-780 89223-782 95020-976
Synthetic Sea Salt, HiMedia Laboratories
Media for Microbiology Additives and Supplements for Microbiology
Synthetic sea salt is recommended for preparation of special diluents.
  • Stored at 2 to 8 °C (prepared medium)
It is used as a diluent during detection and enumeration of E. coli and coliforms in surface and wastewater. Sodium chloride in the medium helps to maintain osmotic
balance and the other ingredients provide buffering action.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty